CAS RN 1313-27-5
Molybdenum(VI) oxide, 99+%
CAS: 1313-27-5 Molecular Formula: MoO3 Molecular Weight (g/mol): 143.95 MDL Number: MFCD00003469 InChI Key: JKQOBWVOAYFWKG-UHFFFAOYSA-N Synonym: molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide PubChem CID: 14802 ChEBI: CHEBI:30627 IUPAC Name: trioxomolybdenum SMILES: O=[Mo](=O)=O
Molybdenum(VI) oxide, Puratronic™, 99.9995% (metals basis excluding W)
CAS: 1313-27-5 Molecular Formula: MoO3 Molecular Weight (g/mol): 143.95 MDL Number: MFCD00003469 InChI Key: JKQOBWVOAYFWKG-UHFFFAOYSA-N Synonym: molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide PubChem CID: 14802 ChEBI: CHEBI:30627 IUPAC Name: trioxomolybdenum SMILES: O=[Mo](=O)=O
Molybdenum(VI) oxide, 99.5%
CAS: 1313-27-5 Molecular Formula: MoO3 Molecular Weight (g/mol): 143.95 MDL Number: MFCD00003469 InChI Key: JKQOBWVOAYFWKG-UHFFFAOYSA-N Synonym: molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide PubChem CID: 14802 ChEBI: CHEBI:30627 IUPAC Name: trioxomolybdenum SMILES: O=[Mo](=O)=O
Molybdenum(VI) oxide, 99.95% (metals basis)
CAS: 1313-27-5 Molecular Formula: MoO3 Molecular Weight (g/mol): 143.95 MDL Number: MFCD00003469 InChI Key: JKQOBWVOAYFWKG-UHFFFAOYSA-N Synonym: molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide PubChem CID: 14802 ChEBI: CHEBI:30627 IUPAC Name: trioxomolybdenum SMILES: O=[Mo](=O)=O
Molybdenum(VI) oxide, 99.5+%, ACS reagent
CAS: 1313-27-5 Molecular Formula: MoO3 Molecular Weight (g/mol): 143.95 MDL Number: MFCD00003469 InChI Key: JKQOBWVOAYFWKG-UHFFFAOYSA-N Synonym: molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide PubChem CID: 14802 ChEBI: CHEBI:30627 IUPAC Name: trioxomolybdenum SMILES: O=[Mo](=O)=O
Molybdenum(VI) oxide, Puratronic™, 99.998% (metals basis excluding W), W 300ppm max
CAS: 1313-27-5 Molecular Formula: MoO3 Molecular Weight (g/mol): 143.95 MDL Number: MFCD00003469 InChI Key: JKQOBWVOAYFWKG-UHFFFAOYSA-N Synonym: molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide PubChem CID: 14802 ChEBI: CHEBI:30627 IUPAC Name: trioxomolybdenum SMILES: O=[Mo](=O)=O
Molybdenum(VI) oxide, ACS, 99.5% min
CAS: 1313-27-5 Molecular Formula: MoO3 Molecular Weight (g/mol): 143.95 MDL Number: MFCD00003469 InChI Key: JKQOBWVOAYFWKG-UHFFFAOYSA-N Synonym: molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide PubChem CID: 14802 ChEBI: CHEBI:30627 IUPAC Name: trioxomolybdenum SMILES: O=[Mo](=O)=O
MP Biomedicals™ Molybdenum Trioxide, ≥99.5%, (ACS Reagent Grade), MP Biomedicals
CAS: 1313-27-5 Molecular Formula: MoO3 Molecular Weight (g/mol): 143.95 MDL Number: MFCD00003469 InChI Key: JKQOBWVOAYFWKG-UHFFFAOYSA-N Synonym: molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide PubChem CID: 14802 ChEBI: CHEBI:30627 IUPAC Name: trioxomolybdenum SMILES: O=[Mo](=O)=O
Molybdenum Trioxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Molybdenum Trioxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Molybdenum Trioxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Molybdenum(VI) Oxide, 99.5%, Solstice
CAS: 1313-27-5 Molecular Formula: MoO3 Molecular Weight (g/mol): 143.95 MDL Number: MFCD00003469 InChI Key: JKQOBWVOAYFWKG-UHFFFAOYSA-N Synonym: molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide PubChem CID: 14802 ChEBI: CHEBI:30627 IUPAC Name: trioxomolybdenum SMILES: O=[Mo](=O)=O
Molybdenum(VI) oxide, ≥99.5%, Reagent Grade, Honeywell™
CAS: 1313-27-5 Molecular Formula: MoO3 Molecular Weight (g/mol): 143.95 MDL Number: MFCD00003469 InChI Key: JKQOBWVOAYFWKG-UHFFFAOYSA-N Synonym: molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide PubChem CID: 14802 ChEBI: CHEBI:30627 IUPAC Name: trioxomolybdenum SMILES: O=[Mo](=O)=O
Molybdenum(VI) Oxide, ≥99.5%, ACS Reagent, Honeywell™ Fluka™
CAS: 1313-27-5 Molecular Formula: MoO3 Molecular Weight (g/mol): 143.95 MDL Number: MFCD00003469 InChI Key: JKQOBWVOAYFWKG-UHFFFAOYSA-N Synonym: molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide PubChem CID: 14802 ChEBI: CHEBI:30627 IUPAC Name: trioxomolybdenum SMILES: O=[Mo](=O)=O