missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum Trioxide, TRC

Product Code. 30470705
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470705 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470705 Supplier Toronto Research Chemicals Supplier No. TRCM4890082.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Molybdenum trioxide
CAS 1313-27-5
Quantity 2.5 g
Molecular Formula MoO3
Synonym Molybdenum oxide (MoO3), Bouen SKN 301, JCPDS 35-0609, MFR, MOP-P 100, Mo 1202T, Molybdena, Molybdenum oxide, Molybdenum oxide (Mo4O12), Molybdenum oxide (Mo5O15), Molybdenum trioxide, Molybdenum trioxide pentamer, Molybdenum trioxide tetramer, Molybdenum(VI) oxide, Molybdenum(VI) trioxide, Molybdic acid anhydride, Molybdic anhydride, Molybdic oxide, NSC 215191, Pol-U, SS-M 0100
InChI Formula InChI=1S/Mo.3O
SMILES O=[Mo](=O)=O
IUPAC Name trioxomolybdenum
Molecular Weight (g/mol) 143.95
Formula Weight 145.8902
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.