accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 131341-86-1

IUPAC Name: 4-(2,2-difluoro-2H-1,3-benzodioxol-4-yl)-1H-pyrrole-3-carbonitrile
Synonyms: fludioxonil
Molecular Weight (g/mol): 248.19
Molecular Formula: C12H6F2N2O2
InChi Key: MUJOIMFVNIBMKC-UHFFFAOYSA-N
SMILES: FC1(F)OC2=CC=CC(=C2O1)C1=CNC=C1C#N
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results