missing translation for 'onlineSavingsMsg'
Learn More

Fludioxonil, TRC

Product Code. 30004073
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004073 5 g 5g
17953315 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004073 Supplier Toronto Research Chemicals Supplier No. TRCF4270005G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131341-86-1
Molecular Formula C12 H6 F2 N2 O2
Molecular Weight (g/mol) 248.18
Synonym 1H-Pyrrole-3-carbonitrile, 4-(2,2-difluoro-1,3-benzodioxol-4-yl)- (9CI, ACI), 4-(2,2-Difluoro-1,3-benzodioxol-4-yl)-1H-pyrrole-3-carbonitrile (ACI), Beret Gold, Celeste, CGA 173506, Fludioxonil, Geoxe, Maxim, Maxim (pesticide), Maxim 4FS, Maxim PSP, Maxin XL, Saphire, Savior, Scholar, Seibia, Spirato, Spirato 480 FS
IUPAC Name 4-(2,2-difluoro-1,3-benzodioxol-4-yl)-1H-pyrrole-3-carbonitrile
SMILES FC1(F)Oc2cccc(c2O1)c3c[nH]cc3C#N

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H6F2N2O2/c13-12(14)17-10-3-1-2-8(11(10)18-12)9-6-16-5-7(9)4-15/h1-3,5-6,16H
Formula Weight 248.0397
Percent Purity >95%
Chemical Name or Material Fludioxonil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.