accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OH2OH2ONiONOOONOO

CAS RN 13138-45-9

IUPAC Name: nickel(2+) hexahydrate dinitrate
Synonyms: nickel(II) nitrate
Molecular Weight (g/mol): 290.79
Molecular Formula: H12N2NiO12
InChi Key: AOPCKOPZYFFEDA-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results