CAS RN 1314-13-2
Zinc oxide, ACS reagent
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, 99.5+%
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, 99.999%, (trace metal basis)
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, Puratronic™, 99.999% (metals basis)
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, 99.9% (metals basis)
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, 99.99% (metals basis)
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, heavy, 98%
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, NanoTek∣r
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, ACS, 99.0% min
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, NanoArc™ ZN-0625, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion with dispersant
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
Zinc oxide, 99.9% (metals basis)
CAS: 1314-13-2 Molecular Formula: OZn Molecular Weight (g/mol): 81.38 MDL Number: MFCD00011300 InChI Key: XLOMVQKBTHCTTD-UHFFFAOYSA-N Synonym: zinc oxide,zinc white,amalox,permanent white,Chinese white,snow white,emanay zinc oxide,felling zinc oxide,zinc oxide zno,akro-zinc bar 85 PubChem CID: 14806 ChEBI: CHEBI:36560 IUPAC Name: oxozinc SMILES: O=[Zn]
- 1
- 2