accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOYbYb

CAS RN 1314-37-0

IUPAC Name: diytterbium(3+) trioxidandiide
Synonyms: ytterbium(III) oxide
Molecular Weight (g/mol): 394.11
Molecular Formula: O3Yb2
InChi Key: UZLYXNNZYFBAQO-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Yb+3].[Yb+3]
Physical Form 
  • (8)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
Grade 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (9)
  • (8)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)