accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1314-98-3

SZn

CAS RN 1314-98-3

IUPAC Name: sulfanylidenezinc
Synonyms: zinc sulfide
Molecular Weight (g/mol): 97.44
Molecular Formula: SZn
InChi Key: WGPCGCOKHWGKJJ-UHFFFAOYSA-N
SMILES: S=[Zn]
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (5)
  • (4)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (6)
  • (6)
  • (1)
Percent Purity 
  • (3)
  • (7)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (15)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (5)
  • (4)
  • (2)
  • (1)
  • (3)
  • (6)
  • (6)
  • (1)
  • (3)
  • (7)
  • (3)