missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfide, Pure, Fisher Chemical

Product Code. 10763811 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10763811

Brand: Fisher Chemical Z/1700/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1314-98-3
Color White
pH 5 to 5.5
Physical Form Solid
Quantity 500 g
Molecular Formula SZn
MDL Number MFCD00011301
Synonym zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
InChI Key DRDVZXDWVBGGMH-UHFFFAOYSA-N
SMILES [S--].[Zn++]
IUPAC Name sulfanylidenezinc
Molecular Weight (g/mol) 97.44
PubChem CID 14821
Formula Weight 97.44g/mol
Packaging Plastic powder jar
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.