accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FeS

CAS RN 1317-37-9

IUPAC Name: sulfanylideneiron
Synonyms: iron(II) sulfide
Molecular Weight (g/mol): 87.91
Molecular Formula: FeS
InChi Key: MBMLMWLHJBBADN-UHFFFAOYSA-N
SMILES: S=[Fe]
Quantity 
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (10)
  • (1)
  • (1)
Grade 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (10)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (10)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)