accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFFFFOBr

CAS RN 131805-94-2

IUPAC Name: 1-[3,5-bis(trifluoromethyl)phenyl]-2-bromoethan-1-one
Synonyms: 1-[3,5-bis(trifluoromethyl)phenyl]-2-bromoethanone
Molecular Weight (g/mol): 335.04
Molecular Formula: C10H5BrF6O
InChi Key: ZEKBFXJTIAEUOF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(=C1)C(=O)CBr)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results