accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFHO

CAS RN 132248-58-9

IUPAC Name: 2,2,2-trifluoroethan-1-ol
Synonyms: trifluoroethanol TFE fluorinol 85
Molecular Weight (g/mol): 100.04
Molecular Formula: C2H3F3O
InChi Key: RHQDFWAXVIIEBN-UHFFFAOYSA-N
SMILES: OCC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results