accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HNSHNNH2

CAS RN 13278-67-6

IUPAC Name: 3-amino-1-(4-methylphenyl)thiourea
Synonyms: 3-amino-1-(4-methylphenyl)thiourea
Molecular Weight (g/mol): 181.26
Molecular Formula: C8H11N3S
InChi Key: IEAWRKQVDLFINI-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC(=S)NN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results