accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13290-96-5

IUPAC Name: 1,3-dimethyl 5-nitrobenzene-1,3-dicarboxylate
Synonyms: 1,3-dimethyl 5-nitrobenzene-1,3-dicarboxylate
Molecular Weight (g/mol): 239.18
Molecular Formula: C10H9NO6
InChi Key: GGTSJKFPGKFLCZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC(=C1)C(=O)OC)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results