CAS RN 133407-82-6
IUPAC Name:
benzyl N-{3-methyl-1-[(3-methyl-1-{[(2S)-4-methyl-1-oxopentan-2-yl]carbamoyl}butyl)carbamoyl]butyl}carbamate
Synonyms:
benzyl N-{3-methyl-1-[(3-methyl-1-{[(2S)-4-methyl-1-oxopentan-2-yl]carbamoyl}butyl)carbamoyl]butyl}carbamate
Molecular Weight (g/mol):
475.63
Molecular Formula:
C26H41N3O5
InChi Key:
TZYWCYJVHRLUCT-UVKLAMSESA-N
SMILES:
CC(C)C[C@H](NC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1)C=O
1
–
5
of
5
results
1
2
MG 132, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
MG 132, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
R&D Systems™ MG 132
A Tocris product, part of R&D Systems. Proteasome and calpain inhibitor. Inhibits NF-κB activation
5
STEMCELL Technologies (S)-MG132
Ubiquitin-proteasome and NF-κB pathway inhibitor; Inhibits proteasome and stabilizes IκB