accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 133454-47-4

CH3OONNOFCH3O

CAS RN 133454-47-4

IUPAC Name: 1-(4-{3-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]propoxy}-3-methoxyphenyl)ethan-1-one
Synonyms: iloperidone zomaril
Molecular Weight (g/mol): 426.49
Molecular Formula: C24H27FN2O4
InChi Key: XMXHEBAFVSFQEX-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1OCCCN1CCC(CC1)C1=NOC2=CC(F)=CC=C12)C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results