accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 134-81-6

IUPAC Name: diphenylethane-1,2-dione
Synonyms: benzil bibenzoyl
Molecular Weight (g/mol): 210.23
Molecular Formula: C14H10O2
InChi Key: WURBFLDFSFBTLW-UHFFFAOYSA-N
SMILES: O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)