accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(S)OOCH3CH3H3CNH2HCl

CAS RN 13404-22-3

IUPAC Name: tert-butyl 2-aminopropanoate hydrochloride
Synonyms: tert-butyl 2-aminopropanoate hydrochloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: WIQIWPPQGWGVHD-UHFFFAOYNA-N
SMILES: Cl.CC(N)C(=O)OC(C)(C)C
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results