missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine tert-Butyl Ester Hydrochloride, TRC

Product Code. 30310248
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310248 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310248 Supplier Toronto Research Chemicals Supplier No. TRCA4810155G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Alanine tert-Butyl Ester Hydrochloride
CAS 13404-22-3
Molecular Formula C7H16ClNO2
Quantity 5 g
Synonym L-Alanine 1,1-Dimethylethyl Ester Hydrochloride, (S)-2-Aminopropionic Acid tert-Butyl Ester Hydrochloride, (S)-Alanine tert-Butyl Ester Hydrochloride, L-Alanine tert-Butyl Ester Hydrochloride, tert-Butyl (S)-2-Aminopropanoate Hydrochloride, tert-Butyl L-Alaninate Hydrochloride
InChI Formula InChI=1S/C7H15NO2.ClH/c1-5(8)6(9)10-7(2,3)4;/h5H,8H2,1-4H3;1H/t5-;/m0./s1
SMILES Cl.C[C@H](N)C(=O)OC(C)(C)C
IUPAC Name tert-butyl (2S)-2-aminopropanoate;hydrochloride
Molecular Weight (g/mol) 181.66
Formula Weight 181.087
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.