accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13414-95-4

OS

CAS RN 13414-95-4

IUPAC Name: 4,5,6,7-tetrahydro-1-benzothiophen-4-one
Synonyms: 6,7-dihydro-5H-1-benzothiophen-4-one
Molecular Weight (g/mol): 152.21
Molecular Formula: C8H8OS
InChi Key: GJEKNELSXNSYAQ-UHFFFAOYSA-N
SMILES: O=C1CCCC2=C1C=CS2
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)