accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOONCO

CAS RN 13471-69-7

IUPAC Name: 4-isocyanato-1-methyl-2-nitrobenzene
Synonyms: 4-isocyanato-1-methyl-2-nitrobenzene
Molecular Weight (g/mol): 178.15
Molecular Formula: C8H6N2O3
InChi Key: OIORBBLUSMONPW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1[N+]([O-])=O)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results