Learn More
4-Methyl-3-nitrophenyl isocyanate, 96%, Thermo Scientific™
Product Code. 11313306
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313306 5 g 5g
11303306 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11313306 Supplier Thermo Scientific Alfa Aesar Supplier No. L12669.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13471-69-7
Molecular Formula C8H6N2O3
Molecular Weight (g/mol) 178.147
MDL Number MFCD00014698
InChI Key OIORBBLUSMONPW-UHFFFAOYSA-N
PubChem CID 73137
IUPAC Name 4-isocyanato-1-methyl-2-nitrobenzene
SMILES CC1=C(C=C(C=C1)N=C=O)[N+](=O)[O-]
Specifications
Melting Point 48°C to 54°C
Boiling Point 113°C to 118°C (3 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Quantity 5 g
UN Number UN2811
Beilstein 2724128
Sensitivity Moisture sensitive
Synonym 4-methyl-3-nitrophenyl isocyanate, 2-nitro-4-isocyanatotoluene, 4-methyl-3-nitrophenylisocyanate, benzene, 4-isocyanato-1-methyl-2-nitro, benzene,4-isocyanato-1-methyl-2-nitro, 4-methyl-3-nitrobenzenisocyanate, acmc-20amdt
Formula Weight 178.15
Percent Purity 96%
Chemical Name or Material 4-Methyl-3-nitrophenyl isocyanate