accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OOHPOOHONa

CAS RN 13472-35-0

IUPAC Name: sodium dihydrate dihydrogen phosphate
Synonyms: sodium dihydrate dihydrogen phosphate
Molecular Weight (g/mol): 156.01
Molecular Formula: H6NaO6P
InChi Key: VBJGJHBYWREJQD-UHFFFAOYSA-M
SMILES: O.O.[Na+].OP(O)([O-])=O
Molecular Weight (g/mol) 
  • (18)
  • (13)
Quantity 
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (6)
  • (1)
Percent Purity 
  • (1)
  • (4)
  • (5)
  • (3)
  • (1)
  • (4)
Physical Form 
  • (3)
  • (2)
  • (1)
  • (9)
Grade 
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (9)
  • (5)
  • (2)
  • (2)
  • (7)
  • (4)
  • (2)
  • (4)
  • (3)
  • (18)
  • (13)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (6)
  • (1)
  • (1)
  • (4)
  • (5)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (9)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)