accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 135-73-9

BrO

CAS RN 135-73-9

IUPAC Name: 1-{[1,1'-biphenyl]-4-yl}-2-bromoethan-1-one
Synonyms: 1-{[1,1'-biphenyl]-4-yl}-2-bromoethanone
Molecular Weight (g/mol): 275.15
Molecular Formula: C14H11BrO
InChi Key: KGHGZRVXCKCJGX-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)