accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13506-76-8

IUPAC Name: 2-methyl-6-nitrobenzoic acid
Synonyms: 2-methyl-6-nitrobenzoic acid
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: CCXSGQZMYLXTOI-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1C(O)=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results