accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3COH3CF

CAS RN 1351499-39-2

IUPAC Name: 1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-one
Synonyms: 1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
Molecular Weight (g/mol): 264.1
Molecular Formula: C14H18BFO3
InChi Key: UDNSDVWJDRQRHR-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C=C(C=C1)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)