accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13528-93-3

IUPAC Name: chloro[2-(chlorodimethylsilyl)ethyl]dimethylsilane
Synonyms: chloro[2-(chlorodimethylsilyl)ethyl]dimethylsilane
Molecular Weight (g/mol): 215.26
Molecular Formula: C6H16Cl2Si2
InChi Key: VGQOKOYKFDUPPJ-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CC[Si](C)(C)Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results