missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(chlorodimethylsilyl)ethane, tech. 90%

Product Code. 11387745
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387745 100 g 100g
11377745 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387745 Supplier Thermo Scientific Alfa Aesar Supplier No. L02570.22
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13528-93-3 | C6H16Cl2Si2 | 215.26 g/mol

1,2-Bis(chlorodimethylsilyl)ethane is used as Protecting Reagent for Primary Amines. It is also used in Medicine field, electronics industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(chlorodimethylsilyl)ethane is used as Protecting Reagent for Primary Amines. It is also used in Medicine field, electronics industry and polymer material.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Incompatible with Strong oxidizing agents, Strong bases and water. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13528-93-3
Molecular Formula C6H16Cl2Si2
Molecular Weight (g/mol) 215.26
MDL Number MFCD00000505
InChI Key VGQOKOYKFDUPPJ-UHFFFAOYSA-N
Synonym 1,2-bis chlorodimethylsilyl ethane, ethylenebis chlorodimethylsilane, silane, 1,2-ethanediylbis chlorodimethyl, unii-b4ocj0e6j1, 1,1,4,4-tetramethyl-1,4-dichlorodisilethylene, chloro 2-chlorodimethylsilyl ethyl dimethylsilane, b4ocj0e6j1, 2,5-dichloro-2,5-dimethyl-2,5-disilahexane, 1,1,4,4-tetramethyl-1,4-dichloro-disylethylene
PubChem CID 83552
SMILES C[Si](C)(Cl)CC[Si](C)(C)Cl

Specifications

Boiling Point 198°C to 200°C
Melting Point 34°C to 40°C
Quantity 100 g
UN Number UN3261
Beilstein 1920141
Flash Point 65°C (149°F)
Solubility Information Reacts with water.
IUPAC Name chloro-[2-[chloro(dimethyl)silyl]ethyl]-dimethylsilane
Formula Weight 215.27
Percent Purity 90%
Grade Technical
Sensitivity Moisture sensitive
Chemical Name or Material 1,2-Bis(chlorodimethylsilyl)ethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.