CAS RN 136470-78-5
IUPAC Name:
[(1S,4R)-4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]cyclopent-2-en-1-yl]methanol
Synonyms:
abacavir ziagen
Molecular Weight (g/mol):
286.34
Molecular Formula:
C14H18N6O
InChi Key:
MCGSCOLBFJQGHM-SCZZXKLOSA-N
SMILES:
NC1=NC(NC2CC2)=C2N=CN([C@@H]3C[C@H](CO)C=C3)C2=N1
1
–
2
of
2
results
2
Abacavir, TRC
CAS: 136470-78-5 Molecular Formula: C14 H18 N6 O Synonym: Abacavir∣2-Cyclopentene-1-methanol, 4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]-, (1S,4R)-∣2-Cyclopentene-1-methanol, 4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]-, (1S-cis)-∣(1S,4R)-4-[2-Amino-6-(cyclopropylamino)-9H-purin-9-yl]-2-cyclopentene-1-methanol∣1592U89∣ABC