accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13679-75-9

CH3OS

CAS RN 13679-75-9

IUPAC Name: 1-(thiophen-2-yl)propan-1-one
Synonyms: 1-(thiophen-2-yl)propan-1-one
Molecular Weight (g/mol): 140.2
Molecular Formula: C7H8OS
InChi Key: MFPZQZZWAMAHOY-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results