accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COPOCH3Cl

CAS RN 13685-30-8

IUPAC Name: chlorobis(4-methoxyphenyl)phosphane
Synonyms: chlorobis(4-methoxyphenyl)phosphane
Molecular Weight (g/mol): 280.69
Molecular Formula: C14H14ClO2P
InChi Key: YTFQUBRFOJIJOZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)P(Cl)C1=CC=C(OC)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results