accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOHOOHOOH

CAS RN 137-66-6

IUPAC Name: 2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate
Synonyms: ascorbyl palmitate
Molecular Weight (g/mol): 414.54
Molecular Formula: C22H38O7
InChi Key: NULLRXHRYOXSEW-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (6)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)