CAS RN 13708-12-8
IUPAC Name:
5-methylquinoxaline
Synonyms:
5-methylquinoxaline
Molecular Weight (g/mol):
144.18
Molecular Formula:
C9H8N2
InChi Key:
CQLOYHZZZCWHSG-UHFFFAOYSA-N
SMILES:
CC1=C2N=CC=NC2=CC=C1
1
–
1
of
1
results
1
5-Methylquinoxaline, 98%
CAS: 13708-12-8 Molecular Formula: C9H8N2 Molecular Weight (g/mol): 144.177 MDL Number: MFCD00012335 InChI Key: CQLOYHZZZCWHSG-UHFFFAOYSA-N Synonym: quinoxaline, 5-methyl,unii-l0ad7t152g,5-methyl quinoxaline,fema no. 3203,8-methylquinoxaline,5-methyl-quinoxaline,acmc-209caq,5-methylquinoxaline,dsstox_cid_27090 PubChem CID: 61670 IUPAC Name: 5-methylquinoxaline SMILES: CC1=C2C(=CC=C1)N=CC=N2