Learn More
5-Methylquinoxaline, 98%
Product Code. 11469556
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469556 5 g 5g
11459556 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11469556 Supplier Thermo Scientific Alfa Aesar Supplier No. A19300.06
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
CAS: 13708-12-8 | C9H8N2 | 144.177 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13708-12-8
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00012335
InChI Key CQLOYHZZZCWHSG-UHFFFAOYSA-N
Synonym quinoxaline, 5-methyl, unii-l0ad7t152g, 5-methyl quinoxaline, fema no. 3203, 8-methylquinoxaline, 5-methyl-quinoxaline, acmc-209caq, 5-methylquinoxaline, dsstox_cid_27090
PubChem CID 61670
IUPAC Name 5-methylquinoxaline
SMILES CC1=C2C(=CC=C1)N=CC=N2
Specifications
Melting Point 20°C to 21°C
Density 1.12
Boiling Point 119°C to 121°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.62
Quantity 5 g
Formula Weight 144.18
Percent Purity 98%
Chemical Name or Material 5-Methylquinoxaline