accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1373920-93-4

H3COFOF

CAS RN 1373920-93-4

IUPAC Name: 4-butoxy-2,6-difluorobenzaldehyde
Synonyms: 4-butoxy-2,6-difluorobenzaldehyde
Molecular Weight (g/mol): 214.21
Molecular Formula: C11H12F2O2
InChi Key: VNPPSJNGJUIJDV-UHFFFAOYSA-N
SMILES: CCCCOC1=CC(F)=C(C=O)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results