CAS RN 13755-38-9
Sodium nitroferricyanide dihydrate, reagent ACS,meets the requirements of Reag.Ph.Eur.
CAS: 13755-38-9 Molecular Formula: C5H4FeN6Na2O3 Molecular Weight (g/mol): 297.95 MDL Number: MFCD00149192 InChI Key: JFPDVRGGFBUORC-UHFFFAOYSA-N Synonym: nitroprusside sodium,sodium pentacyanonitrosylferrate,sodium nitroferricyanide iii dihydrate,dsstox_cid_21126,dsstox_rid_79630,dsstox_gsid_41126,sodium pentacyanonitrosylferrate iii dihydrate,nitropress tn,sodium nitroprusside usp,c5fen6o.2na.2h2o PubChem CID: 11953895 SMILES: O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
Sodium Nitroprusside Dihydrate, Certified AR for Analysis, Fisher Chemical
Sodium nitroferricyanide dihydrate, 99+%
CAS: 13755-38-9 Molecular Formula: C5H4FeN6Na2O3 Molecular Weight (g/mol): 297.95 MDL Number: MFCD00149192 InChI Key: JFPDVRGGFBUORC-UHFFFAOYSA-N Synonym: nitroprusside sodium,sodium pentacyanonitrosylferrate,sodium nitroferricyanide iii dihydrate,dsstox_cid_21126,dsstox_rid_79630,dsstox_gsid_41126,sodium pentacyanonitrosylferrate iii dihydrate,nitropress tn,sodium nitroprusside usp,c5fen6o.2na.2h2o PubChem CID: 11953895 SMILES: O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%
CAS: 13755-38-9 Molecular Formula: C5H4FeN6Na2O3 Molecular Weight (g/mol): 297.95 MDL Number: MFCD00149192 InChI Key: JFPDVRGGFBUORC-UHFFFAOYSA-N Synonym: nitroprusside sodium,sodium pentacyanonitrosylferrate,sodium nitroferricyanide iii dihydrate,dsstox_cid_21126,dsstox_rid_79630,dsstox_gsid_41126,sodium pentacyanonitrosylferrate iii dihydrate,nitropress tn,sodium nitroprusside usp,c5fen6o.2na.2h2o PubChem CID: 11953895 IUPAC Name: disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate SMILES: O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
Sodium pentacyanonitrosylferrate(III) dihydrate, ACS, 99.0-102.0%
CAS: 13755-38-9 Molecular Formula: C5H4FeN6Na2O3 Molecular Weight (g/mol): 297.95 MDL Number: MFCD00149192 InChI Key: JFPDVRGGFBUORC-UHFFFAOYSA-N Synonym: nitroprusside sodium,sodium pentacyanonitrosylferrate,sodium nitroferricyanide iii dihydrate,dsstox_cid_21126,dsstox_rid_79630,dsstox_gsid_41126,sodium pentacyanonitrosylferrate iii dihydrate,nitropress tn,sodium nitroprusside usp,c5fen6o.2na.2h2o PubChem CID: 11953895 IUPAC Name: disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate SMILES: O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
Sodium Nitroprusside Dihydrate, (Crystalline),MP Biomedicals
CAS: 13755-38-9 Molecular Formula: C5H4FeN6Na2O3 Molecular Weight (g/mol): 297.95 MDL Number: MFCD00149192 InChI Key: JFPDVRGGFBUORC-UHFFFAOYSA-N Synonym: nitroprusside sodium,sodium pentacyanonitrosylferrate,sodium nitroferricyanide iii dihydrate,dsstox_cid_21126,dsstox_rid_79630,dsstox_gsid_41126,sodium pentacyanonitrosylferrate iii dihydrate,nitropress tn,sodium nitroprusside usp,c5fen6o.2na.2h2o PubChem CID: 11953895 SMILES: O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
Sodium Nitroprusside Dihydrate, puriss. p.a., ≥99%, ACS reagent, Solstice
CAS: 13755-38-9 Molecular Formula: C5H4FeN6Na2O3 Molecular Weight (g/mol): 297.95 MDL Number: MFCD00149192 InChI Key: JFPDVRGGFBUORC-UHFFFAOYSA-N Synonym: nitroprusside sodium,sodium pentacyanonitrosylferrate,sodium nitroferricyanide iii dihydrate,dsstox_cid_21126,dsstox_rid_79630,dsstox_gsid_41126,sodium pentacyanonitrosylferrate iii dihydrate,nitropress tn,sodium nitroprusside usp,c5fen6o.2na.2h2o PubChem CID: 11953895 IUPAC Name: disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate SMILES: O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
Sodium Nitroprusside Dihydrate, TRC
CAS: 13755-38-9 Molecular Formula: C5 Fe N6 O . 2 Na . 2 H2 O Molecular Weight (g/mol): 297.95 Synonym: Sodium nitroprusside,Sodium pentacyanonitrosylferrate (III) dihydrate,(OC-6-22)-pentakis(cyano-κC)nitrosyl-Ferrate(2-) sodium hydrate (1:2:2),Ferrate(2-), pentacyanonitrosyl-, disodium, dihydrate (8CI),Ferrate(2-), pentakis(cyano-C)nitrosyl-, disodium, dihydrate, (OC-6-22)-,Disodium nitroprusside dihydrate,Disodium pentacyanonitrosylferrate(2-) dihydrate,SNP,Sodium nitroprusside dihydrate IUPAC Name: disodium;pentakis(azanylidynemethyl)-(oxoniumylidyne-λ^{4}-azanyl)iron(2+);dihydrate SMILES: O.O.[Na+].[Na+].N#[C-][Fe+2]([C-]#N)([C-]#N)([C-]#N)([C-]#N)N#[O+]
Sodium Nitroferricyanide(Iii) Dihydrate, Solstice
CAS: 13755-38-9 Molecular Formula: C5H4FeN6Na2O3 Molecular Weight (g/mol): 297.95 MDL Number: MFCD00149192 InChI Key: JFPDVRGGFBUORC-UHFFFAOYSA-N Synonym: nitroprusside sodium,sodium pentacyanonitrosylferrate,sodium nitroferricyanide iii dihydrate,dsstox_cid_21126,dsstox_rid_79630,dsstox_gsid_41126,sodium pentacyanonitrosylferrate iii dihydrate,nitropress tn,sodium nitroprusside usp,c5fen6o.2na.2h2o PubChem CID: 11953895 SMILES: O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
Sodium Nitroprusside Dihydrate, purum p.a., ≥98% (AT), ACS reagent, Honeywell™
CAS: 13755-38-9 Molecular Formula: C5H4FeN6Na2O3 Molecular Weight (g/mol): 297.95 MDL Number: MFCD00149192 InChI Key: JFPDVRGGFBUORC-UHFFFAOYSA-N Synonym: nitroprusside sodium,sodium pentacyanonitrosylferrate,sodium nitroferricyanide iii dihydrate,dsstox_cid_21126,dsstox_rid_79630,dsstox_gsid_41126,sodium pentacyanonitrosylferrate iii dihydrate,nitropress tn,sodium nitroprusside usp,c5fen6o.2na.2h2o PubChem CID: 11953895 SMILES: O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
Sodium Nitroferricyanide, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 13755-38-9 Molecular Formula: C5H4FeN6Na2O3 Molecular Weight (g/mol): 297.95 MDL Number: MFCD00149192 InChI Key: JFPDVRGGFBUORC-UHFFFAOYSA-N Synonym: nitroprusside sodium,sodium pentacyanonitrosylferrate,sodium nitroferricyanide iii dihydrate,dsstox_cid_21126,dsstox_rid_79630,dsstox_gsid_41126,sodium pentacyanonitrosylferrate iii dihydrate,nitropress tn,sodium nitroprusside usp,c5fen6o.2na.2h2o PubChem CID: 11953895 SMILES: O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]