missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroprusside Dihydrate, TRC

Product Code. 30136087
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136087 50 g 50g
30011336 10 g 10g
30012239 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136087 Supplier Toronto Research Chemicals Supplier No. TRCS64802050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5 Fe N6 O . 2 Na . 2 H2 O
Molecular Weight (g/mol) 297.95
Synonym Sodium nitroprusside, Sodium pentacyanonitrosylferrate (III) dihydrate, (OC-6-22)-pentakis(cyano-κC)nitrosyl-Ferrate(2-) sodium hydrate (1:2:2), Ferrate(2-), pentacyanonitrosyl-, disodium, dihydrate (8CI), Ferrate(2-), pentakis(cyano-C)nitrosyl-, disodium, dihydrate, (OC-6-22)-, Disodium nitroprusside dihydrate, Disodium pentacyanonitrosylferrate(2-) dihydrate, SNP, Sodium nitroprusside dihydrate
IUPAC Name disodium;pentakis(azanylidynemethyl)-(oxoniumylidyne-λ^{4}-azanyl)iron(2+);dihydrate
SMILES O.O.[Na+].[Na+].N#[C-][Fe+2]([C-]#N)([C-]#N)([C-]#N)([C-]#N)N#[O+]

Specifications

Quantity 50 g
InChI Formula InChI=1S/5CN.Fe.NO.2Na.2H2O/c5*1-2;;1-2;;;;/h;;;;;;;;;2*1H2/q5*-1;+2;3*+1;;
Formula Weight 297.949
Chemical Name or Material Sodium Nitroprusside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.