accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13991-08-7

PP

CAS RN 13991-08-7

IUPAC Name: [2-(diphenylphosphanyl)phenyl]diphenylphosphane
Synonyms: [2-(diphenylphosphanyl)phenyl]diphenylphosphane
Molecular Weight (g/mol): 446.47
Molecular Formula: C30H24P2
InChi Key: NFRYVRNCDXULEX-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results