missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(diphenylphosphino)benzene, 98%

Product Code. 11464567
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
This item is not returnable. View return policy

Product Code. 11464567

Brand: Thermo Scientific Alfa Aesar B21106.01

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 13991-08-7 | C30H24P2 | 446.47 g/mol

It acts as a ligand for Baeyer-Villiger Oxidation, cycloadditions and reductions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as a ligand for Baeyer-Villiger Oxidation, cycloadditions and reductions.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13991-08-7
Molecular Formula C30H24P2
Molecular Weight (g/mol) 446.47
MDL Number MFCD00014081
InChI Key NFRYVRNCDXULEX-UHFFFAOYSA-N
Synonym 1,2-bis diphenylphosphino benzene, dppbz, dppbe, dppben, dppbenz, o-bis diphenylphosphino benzene, 1,2-bis dimethylphosphino benzene, o-phenylenebis diphenylphosphine, phosphine, phenylenebis diphenyl, 1,2-bis diphenylphosphanyl benzene
PubChem CID 498379
IUPAC Name (2-diphenylphosphanylphenyl)-diphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 185°C to 189°C
Quantity 0.25 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 446.47
Percent Purity 98%
Chemical Name or Material 1,2-Bis(diphenylphosphino)benzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.