accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONHHNNHHN

CAS RN 140-22-7

IUPAC Name: 1,3-bis(phenylamino)urea
Synonyms: 1,5-diphenylcarbazide
Molecular Weight (g/mol): 242.28
Molecular Formula: C13H14N4O
InChi Key: KSPIHGBHKVISFI-UHFFFAOYSA-N
SMILES: O=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (8)
Quantity 
  • (3)
  • (5)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (1)
  • (3)
  • (1)
Grade 
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (8)
  • (3)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)