missing translation for 'onlineSavingsMsg'
Learn More

1,5-DipHenylcarbazide, Extra Pure, SLR, Fisher Chemical

Product Code. 10365940 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10365940 25 g Plastic powder jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10365940 Supplier Fisher Chemical Supplier No. D/4750/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 140-22-7
Melting Point 173°C
Color Beige
Packaging Plastic powder jar
Molecular Formula C13H14N4O
MDL Number 3013
Quantity 25 g
Synonym 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
IUPAC Name 1,3-dianilinourea
Molecular Weight (g/mol) 242.282
PubChem CID 8789
ChEBI CHEBI:4641
Formula Weight 242.28g/mol
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • 1,5-Diphenylcarbazide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.