accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N

CAS RN 140-29-4

IUPAC Name: 2-phenylacetonitrile
Synonyms: phenylacetonitrile novaminsulfon algopyrin benzyl nitrile methamizole novaminsulfone noraminophenazone C13H16N3naO4S natrium novaminsulfonicum novamidazophenum
Molecular Weight (g/mol): 117.15
Molecular Formula: C8H7N
InChi Key: SUSQOBVLVYHIEX-UHFFFAOYSA-N
SMILES: N#CCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results