accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14014-06-3

OHNa

CAS RN 14014-06-3

IUPAC Name: sodium hydroxide
Synonyms: sodium hydroxide NaOH natriumhydroxid soude caustique sodium hydrate hydroxyde de sodium aetznatron soda lye
Molecular Weight (g/mol): 40
Molecular Formula: HNaO
InChi Key: HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES: [OH-].[Na+]
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results