accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 140148-70-5

IUPAC Name: 1-[(tert-butoxy)carbonyl]pyrrolidine-3-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylic acid
Molecular Weight (g/mol): 215.25
Molecular Formula: C10H17NO4
InChi Key: HRMRQBJUFWFQLX-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCC(C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results