missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-beta-proline, 95%, Thermo Scientific Chemicals

Product Code. 15414288
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414288 1 g 1g
15424288 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414288 Supplier Thermo Scientific Alfa Aesar Supplier No. H57888.03
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 140148-70-5 | C10H17NO4 | 215.249 g/mol

N-Boc-L-beta-proline is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-beta-proline is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (slightly).

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140148-70-5
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD03094728
InChI Key HRMRQBJUFWFQLX-ZETCQYMHSA-N
Synonym s-1-boc-pyrrolidine-3-carboxylic acid, s-1-n-boc-beta-proline, s-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, n-boc-l-beta-proline, 3s-1-tert-butoxycarbonyl-3-pyrrolidinecarboxylic acid, s-n-boc-pyrrolidine-3-carboxylic acid, 3s-boc-beta-pro-oh, 3s-1-boc-pyrrolidine-3-carboxylic acid, boc-3s-1-pyrrolidine-3-carboxylic acid
PubChem CID 1512492
IUPAC Name (3S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCC(C1)C(=O)O

Specifications

Quantity 1 g
Solubility Information Soluble in water (slightly).
Optical Rotation +15° (c=0.8 in Chloroform)
Formula Weight 215.25
Percent Purity 95%
Chemical Name or Material N-Boc-L-beta-proline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.