CAS RN 1402222-66-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1R,2S)-2-(3,4-Difluorophenyl)cyclopropanamine Hydrochloride, TRC
CAS: 1402222-66-5 Molecular Formula: C9 H9 F2 N . H Cl Molecular Weight (g/mol): 205.632 Synonym: Ticagrelor Related Compound 31 HCl IUPAC Name: (1R,2S)-2-(3,4-difluorophenyl)cyclopropan-1-amine;hydrochloride SMILES: Cl.N[C@@H]1C[C@H]1c2ccc(F)c(F)c2