missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-2-(3,4-Difluorophenyl)cyclopropanamine Hydrochloride, TRC

Product Code. 18073120
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18073120 5 g 5g
18031850 1 g 1g
18023730 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18073120 Supplier Toronto Research Chemicals Supplier No. TRCD4470305G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1402222-66-5
Molecular Formula C9 H9 F2 N . H Cl
Molecular Weight (g/mol) 205.632
Synonym Ticagrelor Related Compound 31 HCl
IUPAC Name (1R,2S)-2-(3,4-difluorophenyl)cyclopropan-1-amine;hydrochloride
SMILES Cl.N[C@@H]1C[C@H]1c2ccc(F)c(F)c2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H9F2N.ClH/c10-7-2-1-5(3-8(7)11)6-4-9(6)12;/h1-3,6,9H,4,12H2;1H/t6-,9+;/m0./s1
Formula Weight 205.047
Chemical Name or Material (1R,2S)-2-(3,4-Difluorophenyl)cyclopropanamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.