accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHH3CH3CCH3CH3H3CCH3

CAS RN 14035-33-7

IUPAC Name: 1-(3,5-di-tert-butyl-4-hydroxyphenyl)ethan-1-one
Synonyms: 1-(3,5-di-tert-butyl-4-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 248.37
Molecular Formula: C16H24O2
InChi Key: WGJPGMJLARWHRK-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)