accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1405-10-3

ONH2HOHONH2(S)(R)(S)O(R)OHH2NONH2OONH2HOHONH2OHOHOOHSOHOO

CAS RN 1405-10-3

IUPAC Name: (2R,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2-{[(2S,3R,4S,5R)-4-{[(3R,4R,5S,6S)-3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy}-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy}-3-hydroxycyclohexyl]oxy}oxane-3,4-diol; sulfuric acid
Synonyms: (2R,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2-{[(2S,3R,4S,5R)-4-{[(3R,4R,5S,6S)-3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy}-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy}-3-hydroxycyclohexyl]oxy}oxane-3,4-diol; sulfuric acid
Molecular Weight (g/mol): 712.72
Molecular Formula: C23H48N6O17S
InChi Key: OIXVKQDWLFHVGR-GQTDVWSESA-N
SMILES: OS(O)(=O)=O.NC[C@@H]1OC(O[C@@H]2[C@@H](CO)O[C@@H](O[C@@H]3[C@@H](O)[C@H](N)C[C@H](N)[C@H]3O[C@H]3O[C@H](CN)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)[C@H](N)[C@@H](O)[C@@H]1O
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (6)
  • (2)
Color 
  • (2)
  • (1)
  • (5)
Grade 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (5)
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)