missing translation for 'onlineSavingsMsg'
Learn More

Neomycin Sulfate, Phytopure™, MP Biomedicals™

Product Code. 17610907 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Product Code. 17610907

Brand: MP Biomedicals 0219453325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Synonym: Neomycin B Sulfate, Bykomycin, Endomixin, Fraquinol, Myacine, Neosulf, Neomix

Neomycin sulfate is an aminoglycoside antibiotic active against gram-negative bacteria. It is commonly used in many topical medications, such as creams, ointments, and eyedrops. It binds to duplex RNA with high affinity and inhibits protein biosynthesis. Neomycin sulfate also acts as a calcium channel blocker. Usage Statement: Unless specified otherwise, MP Biomedical™ products are for research or further manufacturing use only, not for direct human use.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-10-3

Specifications

Physical Form Powder
Color Light Yellow
Quantity 25 g
compliance-icons compliance-icons
Product Identifier
  • Neomycin Sulphate, Cell Culture Reagent
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Neomycin Sulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.